Learn More
Aviva Systems Biology Corp TFPT Antibody - C-terminal region (ARP65110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFPT. It was validated on Western Blot. . Alternate names: FB1, INO80F, amidaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-432