Learn More
Aviva Systems Biology Corp TFRC Antibody - N-terminal region (ARP63718_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63718_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFRC. It was validated on Western Blot. . Alternate names: CD71, TFR, TFR1, TRFR, T9, TR, p90ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-288-062