Learn More
Electron Microscopy Sciences Cassette Holder TFZ40 & Tray for 40 Cassettes
SDP
Catalog No. 5030560
Encompass
Holds cassettes in a single horizontal plane, which conforms to research regarding the way to achieve the best distribution of microwave energy. It is based upon an original design in wood (the 'Bamboozle') developed by Dr. Richard Dapson of Anatech, Ltd. It is adapted to a pyrex container which allows the paraffin to easily hold its temperature, and minimizes the risk of overheating of specimens due to lack of sufficient microwavable load. Holder and tray, holds 40 cassettes in a horizontal plane. Pyrex container allows the paraffin to easily hold its temperature and minimizes the risk of overheating due to lack of sufficient microwavable load.
Catalog No. 50-305-60 Supplier Electron Microscopy Sciences Supplier No. 9707110
Holds cassettes in a single horizontal plane, which conforms to research regarding the way to achieve the best distribution of microwave energy. It is based upon an original design in wood (the 'Bamboozle') developed by Dr. Richard Dapson of Anatech, Ltd. It is adapted to a pyrex container which allows the paraffin to easily hold its temperature, and minimizes the risk of overheating of specimens due to lack of sufficient microwavable load. Holder and tray, holds 40 cassettes in a horizontal plane. Pyrex container allows the paraffin to easily hold its temperature and minimizes the risk of overheating due to lack of sufficient microwavable load.