Learn More
Electron Microscopy Sciences Cassette Holder TFZ64 Only for 64 Cassettes
SDP
Catalog No. 5030563
Encompass
Holds cassettes in a single horizontal plane, which conforms to research regarding the way to achieve the best distribution of microwave energy. It is based upon an original design in wood (the 'Bamboozle') developed by Dr. Richard Dapson of Anatech, Ltd. It is adapted to a pyrex container which allows the paraffin to easily hold its temperature, and minimizes the risk of overheating of specimens due to lack of sufficient microwavable load. Holder only, holds 64 cassettes in a single horizontal plane.
Catalog No. 50-305-63 Supplier Electron Microscopy Sciences Supplier No. 9707002
Holds cassettes in a single horizontal plane, which conforms to research regarding the way to achieve the best distribution of microwave energy. It is based upon an original design in wood (the 'Bamboozle') developed by Dr. Richard Dapson of Anatech, Ltd. It is adapted to a pyrex container which allows the paraffin to easily hold its temperature, and minimizes the risk of overheating of specimens due to lack of sufficient microwavable load. Holder only, holds 64 cassettes in a single horizontal plane.