Learn More
Medchemexpress LLC TG 100572 hydrochloride | 867331-64-4 | 98.8% | 512.43 g/mol | C26H27Cl2N5O2 | 1 ML
SDP
Catalog No. 5000246057
Encompass_Preferred
TG 100572 Hydrochloride is the hydrochloride salt of a multi-targeted kinase inhibitor that blocks selected receptor tyrosine kinases and Src family kinases. It is provided for research use in biochemical and cell-based studies and is available as a ready-to-use solution or as a solid form.
  • Multi-targeted kinase inhibitor active against receptor tyrosine kinases and Src family kinases.
  • Supplied as a 10 mM solution in DMSO (1 mL) for immediate use.
  • Also available as a solid with a pink to red appearance.
  • High purity (~98.8%) suitable for research applications.
  • Store sealed, away from moisture; in solvent store at -80°C for up to 6 months.
Catalog No. 50-002-46057 Supplier Medchemexpress LLC Supplier No. HY1018510MM 1ML
TG 100572 Hydrochloride is the hydrochloride salt of a multi-targeted kinase inhibitor that blocks selected receptor tyrosine kinases and Src family kinases. It is provided for research use in biochemical and cell-based studies and is available as a ready-to-use solution or as a solid form.
  • Multi-targeted kinase inhibitor active against receptor tyrosine kinases and Src family kinases.
  • Supplied as a 10 mM solution in DMSO (1 mL) for immediate use.
  • Also available as a solid with a pink to red appearance.
  • High purity (~98.8%) suitable for research applications.
  • Store sealed, away from moisture; in solvent store at -80°C for up to 6 months.