Learn More
Medchemexpress LLC TG 100572 hydrochloride | 867331-64-4 | MFCD18251422 | 98.8% | 512.43 g/mol | C26H27Cl2N5O2 | 50 MG
SDP
Catalog No. 5000249317
Encompass_Preferred
TG 100572 Hydrochloride is a multi-targeted kinase inhibitor supplied as a hydrochloride salt for laboratory research. It inhibits several receptor tyrosine kinases and Src family kinases and is used in vitro to study signaling pathways and kinase-dependent cellular processes. Offered in multiple pack sizes with high listed purity for biochemical and cell-based studies.
  • Multi-targeted kinase inhibition profile (VEGFR, FGFR, PDGFR, Src family).
  • High listed purity suitable for in vitro assays.
  • Available in small pack sizes and solution format for flexibility.
  • Compatible with biochemical and cell-based assay workflows.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-49317 Supplier Medchemexpress LLC Supplier No. HY1018550MG
TG 100572 Hydrochloride is a multi-targeted kinase inhibitor supplied as a hydrochloride salt for laboratory research. It inhibits several receptor tyrosine kinases and Src family kinases and is used in vitro to study signaling pathways and kinase-dependent cellular processes. Offered in multiple pack sizes with high listed purity for biochemical and cell-based studies.
  • Multi-targeted kinase inhibition profile (VEGFR, FGFR, PDGFR, Src family).
  • High listed purity suitable for in vitro assays.
  • Available in small pack sizes and solution format for flexibility.
  • Compatible with biochemical and cell-based assay workflows.
  • For research use only; not for human or veterinary use.