Learn More
Research Products International Corp TG 10X Solution, 2 L
SDP
Catalog No. 501977995
Encompass_Preferred
10X Formulation:
TRIS Base: 0.25 M
Glycine: 1.92 M
pH: 8.5
Add methanol according to protocol
Catalog No. 50-197-7995 Supplier Research Products International Corp Supplier No. T274002000.0
10X Formulation:
TRIS Base: 0.25 M
Glycine: 1.92 M
pH: 8.5
Add methanol according to protocol