Learn More
Alkali Scientific TG 10X Solution [TRIS Glycine Buffer 10X Solution], 1 Liter
SDP
Catalog No. 5000661625
Encompass_Preferred
TRIS Glycine Buffer 10X Solution is a high-performance buffer designed for use in electrophoresis, particularly for running SDS-PAGE gels. The 1-liter bottle of 10X solution allows for efficient dilution and preparation of multiple electrophoresis runs. This buffer system is well-suited for separating a wide range of proteins, providing reliable and consistent results. The TRIS-Glycine formulation is known for its excellent buffering capacity and is widely used in molecular biology and biochemistry for protein analysis. This buffer is compatible with numerous protein electrophoresis protocols, including those for high-precision applications such as Western blotting and protein quantification.
Catalog No. 50-006-61625 Supplier Alkali Scientific Supplier No. TGX002CP
TRIS Glycine Buffer 10X Solution is a high-performance buffer designed for use in electrophoresis, particularly for running SDS-PAGE gels. The 1-liter bottle of 10X solution allows for efficient dilution and preparation of multiple electrophoresis runs. This buffer system is well-suited for separating a wide range of proteins, providing reliable and consistent results. The TRIS-Glycine formulation is known for its excellent buffering capacity and is widely used in molecular biology and biochemistry for protein analysis. This buffer is compatible with numerous protein electrophoresis protocols, including those for high-precision applications such as Western blotting and protein quantification.