Learn More
Research Products International Corp TG Buffer Powder 10X Powder 1L
SDP
Catalog No. 5000052443
Encompass_Preferred
Dry homogeneous blend of molecular biology grade TRIS base and Glycine. Add 174.4g to make 1L of 10x Concentrate.
Catalog No. 50-000-52443 Supplier Research Products International Corp Supplier No. T290401.0
Dry homogeneous blend of molecular biology grade TRIS base and Glycine. Add 174.4g to make 1L of 10x Concentrate.