Learn More
Bioworld Tris-Glycine-SDS (TG-SDS) Buffer, Ready-Pack (10X), 1 PK(s)
Catalog No. 50196716
Encompass_Preferred
Tris Glycine-SDS BufferTD-SDS Buffer is blended using bioPLUS reagents to provide the highest quality buffer for protein research. Ideal for protein separations, this buffer is formulated to ensure exceptional purity in the absence of nuclease and protease activities.TG-SDS Buffer is a widely used running buffer for denaturing electrophoresis of proteins (SDS-PAGE).Technical Information:Storage: RTHazard: Irritant1 pack makes 1 L of 10x concentrate.1X Solution Contains: 0.025 M Tris, 0.192 M Glycine and 0.1% SDS.
Catalog No. 50-196-716 Supplier Bioworld Supplier No. 105300371
Add to Cart
Add to Cart
Tris Glycine-SDS BufferTD-SDS Buffer is blended using bioPLUS reagents to provide the highest quality buffer for protein research. Ideal for protein separations, this buffer is formulated to ensure exceptional purity in the absence of nuclease and protease activities.TG-SDS Buffer is a widely used running buffer for denaturing electrophoresis of proteins (SDS-PAGE).Technical Information:Storage: RTHazard: Irritant1 pack makes 1 L of 10x concentrate.1X Solution Contains: 0.025 M Tris, 0.192 M Glycine and 0.1% SDS.