Learn More
Medchemexpress LLC HY-15338 10mg Medchemexpress, TG003 CAS:719277-26-6 Purity:>98%
SDP
Catalog No. 502081709
Encompass_Preferred
Medchemexpress, HY-15338 10mg TG003 CAS:719277-26-6 TG003 is a potent inhibitor of Clk1/Sty; inhibits Clk1 and Clk4 with IC50 values of 20 and 15 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1709 Supplier Medchemexpress LLC Supplier No. HY1533810MG
Medchemexpress, HY-15338 10mg TG003 CAS:719277-26-6 TG003 is a potent inhibitor of Clk1/Sty; inhibits Clk1 and Clk4 with IC50 values of 20 and 15 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.