Learn More
Apexbio Technology LLC TG003 300801-52-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062628
Encompass_Preferred
TG003 (CAS 300801-52-9) is a selective inhibitor targeting the Cdc2-like kinase (Clk) family and casein kinase 1 (CK1) Specifically it demonstrates notable potency against mClk1 mClk2 and mClk4 with IC50 values of approximately 20 nM 200 nM and 15 nM respectively TG003 competitively binds with ATP at Clk1 modulating Clk-mediated phosphorylation of serine/arginine-rich (SR) proteins involved in alternative mRNA splicing Studies using HeLa cellular extracts and animal models have shown that TG003 alters splice site selection and pre-mRNA processing suggesting potential utility for investigating RNA splicing regulatory mechanisms
Catalog No. 50-000-62628 Supplier Apexbio Technology LLC Supplier No. B14315.1
TG003 (CAS 300801-52-9) is a selective inhibitor targeting the Cdc2-like kinase (Clk) family and casein kinase 1 (CK1) Specifically it demonstrates notable potency against mClk1 mClk2 and mClk4 with IC50 values of approximately 20 nM 200 nM and 15 nM respectively TG003 competitively binds with ATP at Clk1 modulating Clk-mediated phosphorylation of serine/arginine-rich (SR) proteins involved in alternative mRNA splicing Studies using HeLa cellular extracts and animal models have shown that TG003 alters splice site selection and pre-mRNA processing suggesting potential utility for investigating RNA splicing regulatory mechanisms