Learn More
AdipoGen TG007 (5 mg)
SDP
Supplier:  AdipoGen AGCR13666M005
Encompass_Preferred
Chemical. CAS: 719277-30-2. Formula: C14H15NO3S. MW: 277.3. Synthetic. Prodrug form (acetylated derivative) of INDY (Prod. No. AG-CR1-3665). Cell permeable ATP-competitive DYRK1A and DYRK1B inhibitor. Inhibits phosphorylation of Tau-protein and rescues repressed calcineurin/NFAT signaling. Recovers Xenopus embryos from head malformation induced by DYRK1A overexpression in vivo.
Catalog No. 50-205-4048