Learn More
Agilent Technologies TG1 ELECTROCOMP CELLS
Catalog No. 50125007
Encompass
Cell, TG1 Electroporation-Competent; Stratagene; For phage-display library preparation greater than equal1 x 1010 transformations/ug; F episome for single-strand rescue and blue/white color screening; Highest transformation
Catalog No. 50-125-007 Supplier Agilent Technologies Supplier No. 200123
Cell, TG1 Electroporation-Competent; Stratagene; For phage-display library preparation greater than equal1 x 1010 transformations/ug; F episome for single-strand rescue and blue/white color screening; Highest transformation