Learn More
INTACT GENOMICS INC TG1 Phage Display Electrocompetent Cells 6x100µl
Catalog No. 5000082890
Encompass
Intact Genomics TG1 Phage Display ElectroCompetent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries.Intact Genomics TG1 Phage Display ElectroCompetent Cells have the following features: • ≥4 x 10^10cfu/µg efficiency with electroporation. • Amber suppressor strain (supE)Product Include and Storage: TG1 phage display electrocompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4ºC
Catalog No. 50-000-82890 Supplier INTACT GENOMICS INC Supplier No. 126424
Intact Genomics TG1 Phage Display ElectroCompetent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries.Intact Genomics TG1 Phage Display ElectroCompetent Cells have the following features: • ≥4 x 10^10cfu/µg efficiency with electroporation. • Amber suppressor strain (supE)Product Include and Storage: TG1 phage display electrocompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4ºC