Learn More
AMID BIOSCIENCES Amid Biosciences, TG1 Phage Display Chemically Competent E. coli Cells, 10 x 0.1 mL
SDP
Catalog No. NC4043487
Encompass_Preferred
TG1 phage display chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for convenient transformation during display library preparation and clone handling workflows. The strain is commonly used for filamentous phage display and phagemid propagation. Applications include phage display library preparation, antibody fragment library construction, cloning, and propagation. Technical specifications are included below.- Quantity - 10 x 0.1 mL/tube (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - TG1- Transformation Efficiency - 1.00E+9- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.
Catalog No. NC4043487 Supplier AMID BIOSCIENCES Supplier No. TG120110
TG1 phage display chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for convenient transformation during display library preparation and clone handling workflows. The strain is commonly used for filamentous phage display and phagemid propagation. Applications include phage display library preparation, antibody fragment library construction, cloning, and propagation. Technical specifications are included below.- Quantity - 10 x 0.1 mL/tube (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - TG1- Transformation Efficiency - 1.00E+9- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.