Learn More
AMID BIOSCIENCES Amid Biosciences, TG1 Phage Display Electrocompetent E. coli Cells, 10 x 0.05 mL
SDP
Catalog No. NC4043486
Encompass_Preferred
TG1 phage display electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for high efficiency transformation of antibody peptide or phagemid display libraries. Applications include large antibody or antibody fragment libraries, phage display, library construction and propagation. Technical specifications are included below.- Quantity - 10 x 0.05 mL/tube (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - TG1- Transformation Efficiency - 1.00E+10- Cell Format - 10 x 0.05 mL electrocompetent cellsThis product is for research use only.
Catalog No. NC4043486 Supplier AMID BIOSCIENCES Supplier No. ETG120110
TG1 phage display electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for high efficiency transformation of antibody peptide or phagemid display libraries. Applications include large antibody or antibody fragment libraries, phage display, library construction and propagation. Technical specifications are included below.- Quantity - 10 x 0.05 mL/tube (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - TG1- Transformation Efficiency - 1.00E+10- Cell Format - 10 x 0.05 mL electrocompetent cellsThis product is for research use only.