Learn More
Apexbio Technology LLC TG100-115, 5mg. Cas: 677297-51-7 MFCD: MFCD16038873. PI3Kγ/PI3Kδ inhibitor
SDP
Supplier:  Apexbio Technology LLC A27545
Encompass_Preferred
TG100-115 is an inhibitor of PI3K and PI3K with IC50 values of 83 nM and 235 nM, respectively. TG100-115 shows no obvious activity against PI3K and PI3K [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4762