Learn More
Enzo Life Sciences TG100-115 (5mg). CAS: 677297-51-7
SDP
Catalog No. 502013437
Encompass_Preferred
Alternative name: 6,7-Bis(3-hydroxyphenyl)pteridine-2,4-diamine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3437 Supplier Enzo Life Sciences Supplier No. ENZCHM1390005
Alternative name: 6,7-Bis(3-hydroxyphenyl)pteridine-2,4-diamine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.