Learn More
Selleck Chemical LLC TG100-115 50mg 677297-51-7
SDP
Catalog No. 508465
Encompass_Preferred
TG100-115 is a PI3K?/? inhibitor with IC50 of 83 nM/235 nM, with little effect on PI3K?/?. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-846-5 Supplier Selleck Chemical LLC Supplier No. S135250MG
TG100-115 is a PI3K?/? inhibitor with IC50 of 83 nM/235 nM, with little effect on PI3K?/?. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.