Learn More
Medchemexpress LLC HY-10410 5mg Medchemexpress, TG101209 CAS:936091-14-4 Purity:>98%
SDP
Catalog No. 501968209
Encompass_Preferred
Medchemexpress, HY-10410 5mg TG101209 CAS:936091-14-4 TG101209 is a selective JAK2 inhibitor with IC50 of 6 nM, less potent to Flt3 and RET with IC50 of 25 nM and 17 nM, appr 30-fold selective for JAK2 than JAK3, and sensitive to JAK2V617F and MPLW515L/K mutations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8209 Supplier Medchemexpress LLC Supplier No. HY104105MG
Medchemexpress, HY-10410 5mg TG101209 CAS:936091-14-4 TG101209 is a selective JAK2 inhibitor with IC50 of 6 nM, less potent to Flt3 and RET with IC50 of 25 nM and 17 nM, appr 30-fold selective for JAK2 than JAK3, and sensitive to JAK2V617F and MPLW515L/K mutations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.