Learn More
Cayman Chemical TG11-77hydrochlorIde 10mg
SDP
Catalog No. 502988301
Encompass_Preferred
An EP2 receptor antagonist (KB = 9.7 nM); selective for EP2 over DP1, EP4, and IP receptors (KBs = 7,320, 5,300, and >10,000 nM, respectively); inhibits the expression of genes encoding COX-2, IL-1β, and IL-6 induced by LPS and the EP2 receptor agonist ONO-AE1-259-01 in mouse BV-2 microglia expressing human EP2 receptors at 0.3 and 1 μM
Catalog No. 50-298-8301 Supplier Cayman Chemical Supplier No. 3018810 MG
An EP2 receptor antagonist (KB = 9.7 nM); selective for EP2 over DP1, EP4, and IP receptors (KBs = 7,320, 5,300, and >10,000 nM, respectively); inhibits the expression of genes encoding COX-2, IL-1β, and IL-6 induced by LPS and the EP2 receptor agonist ONO-AE1-259-01 in mouse BV-2 microglia expressing human EP2 receptors at 0.3 and 1 μM