Learn More
Lovibond™ TG268 Gloss Meter
Catalog No. 15398355 Shop All Tintometer Group Products
Change view
Click to view available options
Display:
Touchscreen
Catalog No. Display
15-398-355 Touchscreen
1 options
Catalog No. 15-398-355 Supplier Lovibond™ Supplier No. 403045
Add to Cart
Add to Cart
Request a Quote
The TG268 portable Gloss Meter features selectable 20°, 60° and 85° measurement angles to measure low, medium and high gloss levels on a wide range of materials.
  • Handheld Tri-Gloss Meter designed to give fast, precise and accurate gloss readings
  • Three measurement angles of 20°, 60° and 85°
  • Compare data quickly and easily with data storage capacity of over 5000 data sets
  • 5 inch touchscreen with high resolution, full view display
The ergonomic design, flexibility and intuitive touch screen technology makes the TG 268 ideal for analyzing solid or opaque materials with low, medium or high gloss surfaces. The meter comes with the ability to select 20°, 60° and 85° measurement angles and the choice of multiple measuring modes making it ideal for quality control in the production process of many industries.
  • Handheld and Portable
    Weighing only 350 grams, the TG 268 is the perfect lightweight portable gloss meter. Its compact and ergonomic design offers easy one-handed operation, helping to prevent operator fatigue. Offering the possibility of 1,000 measurements with 8 hours battery life enables the TG 268 to be used anywhere on site for a prolonged period of time.
  • Flexibility
    The TG 268 offers the user high flexibility. With detector angles of 20°/ 60°/ 85° the TG 268 can measure low, medium and high gloss levels on a wide range of materials. To enhance flexibility even further, basic, statistical, and continuous measurement modes are available to ensure the TG 268 can be set perfectly to the user’s needs.
  • Calibration
    Every TG 268 is supplied with a calibration plate and easy to use on screen calibration guide making the whole procedure fast and simple for the user.
Specifications
Battery Life >1000 times (within 8 hours)
Battery Type Rechargeable Lithium-Ion
Display Touchscreen
Disposable Reusable