Learn More
Lovibond™ TG60 Gloss Meter
Catalog No. 15398354 Shop All Tintometer Group Products
Change view
Click to view available options
Data Storage:
1000 measurements
Catalog No. Data Storage
15-398-354 1000 measurements
1 options
Catalog No. 15-398-354 Supplier Lovibond™ Supplier No. 403050
Add to Cart
Add to Cart
The TG60 portable Gloss Meter features a 60° detector angle for fast, precise and accurate gloss reading of materials with medium gloss.
  • Handheld semi-gloss meter designed to give fast, precise and accurate gloss readings
  • 3.5 inch touchscreen with high resolution, full view display
  • Compare data quickly and easily with a data storage capacity of 1000 data sets
The TG 60 is an entry-level handheld gloss meter. The ergonomic design, flexibility and intuitive touch screen technology makes the TG 60 an ideal choice for analyzing solid or opaque solid materials with medium (or semi-gloss) finishes. Using the standard reference angle of 60° the TG 60 is the ideal choice for general gloss measurements.
  • Handheld and Portable
    Weighing only 350 grams, the TG 60 is the perfect lightweight portable semi-gloss meter. Its compact and ergonomic design offers easy one-handed operation, helping to prevent operator fatigue. Offering the possibility of 1,000 measurements with 8 hours battery life enables the TG 60 to be used anywhere on site for a prolonged period of time.
  • Calibration
    Every TG 60 is supplied with a calibration plate and easy to use on screen calibration guide making the whole procedure fast and simple for the user.
Specifications
Battery Life 8 hr.
Data Storage 1000 measurements
Display Touchscreen
Weight (English) 12.3 oz.
Weight (Metric) 350 g