Learn More
TA Instruments TGA Aluminum Sealed Pan Kit (100 Pans, Lids, Tools), TGA 5500/550, Discovery TGA 1st Gen
Catalog No. 5000580743 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA Aluminum Sealed Pan Kit contains 100 aluminum pans, 100 aluminum lids, 30 stainless steel bails, punch alignment tool, crimping die, and crimping tool. Designed for use with Tzero Sample Press (P/N: 901600.901) and older Standard Sample Press. Compatible with TGA 5500/550, Discovery TGA 1st Gen.
Catalog No. 50-005-80743 Supplier TA Instruments Supplier No. 957352.901
Waters TA Instruments TGA Aluminum Sealed Pan Kit contains 100 aluminum pans, 100 aluminum lids, 30 stainless steel bails, punch alignment tool, crimping die, and crimping tool. Designed for use with Tzero Sample Press (P/N: 901600.901) and older Standard Sample Press. Compatible with TGA 5500/550, Discovery TGA 1st Gen.