Learn More
TA Instruments TGA Die Set for Tzero Sample Press
Supplier:  TA Instruments 957540.901
Encompass_Preferred
Waters TA Instruments TGA Die Set is an optional purple die set for encapsulating environmentally sensitive samples in aluminum pans and lids. Requires Aluminum Sealed Pan Kit (P/N: 957352.901) and Waters TA Instruments Tzero Sample Press (P/N: 901600.901).
Catalog No. 50-005-80618