Learn More
TA Instruments TGA Purge Gas Kit, Copper Tubing, Fittings, Metallically Sealed Connection, Discovery TGA, External Purge Gas Supply
Catalog No. 5000580709 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA Purge Gas Kit consists of copper tubing and fittings for a metallically sealed connection of the Discovery TGA to the external purge gas supply.
Catalog No. 50-005-80709 Supplier TA Instruments Supplier No. 956962.901
Waters TA Instruments TGA Purge Gas Kit consists of copper tubing and fittings for a metallically sealed connection of the Discovery TGA to the external purge gas supply.