Learn More
TA Instruments TGA SDT ICTAC Curie Point Reference Materials Kit, Magnetic, for Discovery TGA 550/55, Certified Magnetic Reference Materials, 0.5 Gram Each, 153°C, 357°C, 554°C, 746°C, 931°C, 1116°C, Includes MSDS Sheets and Certificates of Performance
Catalog No. 5000580617 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA/SDT ICTAC Curie Point Reference Materials Kit with Magnet is for the Discovery TGA 550/55. The kit contains ICTAC certified magnetic reference materials for temperature calibration of TGA and SDT instruments. It includes 0.5 gram of each temperature standard: 153°C, 357°C, 554°C, 746°C, 931°C, and 1116°C, along with MSDS sheets and certificates of performance.
Catalog No. 50-005-80617 Supplier TA Instruments Supplier No. 952541.901
Waters TA Instruments TGA/SDT ICTAC Curie Point Reference Materials Kit with Magnet is for the Discovery TGA 550/55. The kit contains ICTAC certified magnetic reference materials for temperature calibration of TGA and SDT instruments. It includes 0.5 gram of each temperature standard: 153°C, 357°C, 554°C, 746°C, 931°C, and 1116°C, along with MSDS sheets and certificates of performance.