Learn More
TA Instruments TGA/SDT Temperature Calibration Kit, Certified Alumel, Nickel Reference Material, ASTM E-1582-93 Test Procedure, Tweezers, Keeper Magnet, Magnet, Case, TGA/SDT Temperature Calibration Kit
Catalog No. 5000580707 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA / SDT Temperature Calibration Kit includes certified alumel and nickel reference materials for temperature calibration. It also includes ASTM E-1582-93 test procedure for TGA temperature calibration, tweezers, keeper magnet, magnet, and case.
Catalog No. 50-005-80707 Supplier TA Instruments Supplier No. 952384.901
Waters TA Instruments TGA / SDT Temperature Calibration Kit includes certified alumel and nickel reference materials for temperature calibration. It also includes ASTM E-1582-93 test procedure for TGA temperature calibration, tweezers, keeper magnet, magnet, and case.