Learn More
TA Instruments TGA Smart-Seal Pan Assembly and Disassembly Fixture, for Convenient Assembly and Disassembly of Tzero Pan in Smart-Seal Bail Assembly
Catalog No. 5000580636 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA Smart-Seal Pan Assembly and Disassembly Fixture provides a convenient way to assemble and disassemble the Tzero pan in the Smart-Seal bail assembly.
Catalog No. 50-005-80636 Supplier TA Instruments Supplier No. 956950.901
Waters TA Instruments TGA Smart-Seal Pan Assembly and Disassembly Fixture provides a convenient way to assemble and disassemble the Tzero pan in the Smart-Seal bail assembly.