Learn More
TA Instruments TGA Smart-Seal Pan Glovebox Handling Tool, for Transfer of Tzero Pans and Aluminum Foils After Taring into Glovebox, Ensures Pan and Foil Pairs Stay Together, Prevents Foils from Being Blown Away by Purge Gas Flow
Catalog No. 5000580675 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA Smart-Seal Pan Glovebox Handling Tool is used for transferring Tzero pans and aluminum foils after taring on the TGA into a glovebox for sample loading and pan sealing. It ensures tared pan and foil pairs stay together and prevents foils from being blown away by the antechamber purge gas flow.
Catalog No. 50-005-80675 Supplier TA Instruments Supplier No. 956943.901
Waters TA Instruments TGA Smart-Seal Pan Glovebox Handling Tool is used for transferring Tzero pans and aluminum foils after taring on the TGA into a glovebox for sample loading and pan sealing. It ensures tared pan and foil pairs stay together and prevents foils from being blown away by the antechamber purge gas flow.