Learn More
TA Instruments TGA Smart-Seal Pan Kit, 15 Smart-Seal Bail Assemblies, 100 Tzero Pans, 100 Aluminum Foils, Fixture for Bail and Pan Assembly, Purge Gas Kit with Metal Tubes, Glovebox Handling Tool, Die Set for Tzero Press, for Use with Discovery TGA 5500, TGA 550, TGA 55
Catalog No. 5000580892 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA Smart-Seal Pan Kit consists of 15 Smart-Seal bail assemblies, 100 Tzero pans, and 100 aluminum foils. It includes a fixture for convenient assembly and disassembly of the bail and pan, a purge gas kit with metal tubes for TGA connection, a glovebox handling tool for transfer of pans and foils after taring into a glovebox, and a die set for a separately sold Tzero press. It is for use with Discovery TGA 5500, TGA 550 with EGA furnace, and TGA 55 with EGA furnace. A separately sold Tzero press is required.
Catalog No. 50-005-80892 Supplier TA Instruments Supplier No. 956960.901
Waters TA Instruments TGA Smart-Seal Pan Kit consists of 15 Smart-Seal bail assemblies, 100 Tzero pans, and 100 aluminum foils. It includes a fixture for convenient assembly and disassembly of the bail and pan, a purge gas kit with metal tubes for TGA connection, a glovebox handling tool for transfer of pans and foils after taring into a glovebox, and a die set for a separately sold Tzero press. It is for use with Discovery TGA 5500, TGA 550 with EGA furnace, and TGA 55 with EGA furnace. A separately sold Tzero press is required.