Learn More
TA Instruments TGA Weight Set Calibrated, Calibrated Weight Set, Discovery TGA, Balance Mechanism, Weight Calibration, TGA Weight Set Calibrated
Catalog No. 5000580688 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TGA Weight Set (Calibrated) is a calibrated weight set used for weight calibration of the Discovery TGA balance mechanism. It ensures accurate and reproducible instrument operation.
Catalog No. 50-005-80688 Supplier TA Instruments Supplier No. 957341.901
Waters TA Instruments TGA Weight Set (Calibrated) is a calibrated weight set used for weight calibration of the Discovery TGA balance mechanism. It ensures accurate and reproducible instrument operation.