Learn More
Perkin Elmer US LLC Ceramic Sample Pans - Qty. 3
Catalog No. 509052216
Encompass_Preferred
These ceramic sample pans are used with the TGA 4000 and STA 6000. The TGA 4000™ features a top loading balance for easy sample loading and unloading. The furnace and balance are isolated from operators to minimize maintenance, and its fast cooling reduces cycle times. The STA 6000™ provides high performance, reliability, and productivity with an advanced innovative sensor technology. The instrument also measures both sample and reference materials simultaneously to yield greater accuracy and higher quality and is designed for both routine and research applications.
Catalog No. 50-905-2216 Supplier Perkin Elmer US LLC Supplier No. N5200040
These ceramic sample pans are used with the TGA 4000 and STA 6000. The TGA 4000™ features a top loading balance for easy sample loading and unloading. The furnace and balance are isolated from operators to minimize maintenance, and its fast cooling reduces cycle times. The STA 6000™ provides high performance, reliability, and productivity with an advanced innovative sensor technology. The instrument also measures both sample and reference materials simultaneously to yield greater accuracy and higher quality and is designed for both routine and research applications.