Learn More
McKesson General Medical Transparent Film Dressing with Pad 3M Tegaderm +Pad Rectangle 3-1/2 X 4 Inch Frame Style Delivery Without Label Sterile, 25/pack
Catalog No. NC1682429
Encompass_Preferred
3M Tegaderm +Pad Film Dressing with Non-Adherent Pad is an all-in-one dressing that provides a sterile, waterproof, viral and bacterial barrier. These dressings consist of a non-adherent absorbent pad bonded to a thin film transparent dressing. Practical for patients – Waterproof film lets patients bathe or shower without removing dressing. Enhances patient comfort – Unique, non-adherent pad will not stick to the wound, allowing normal wound healing with less pain and trauma. Unique, non-adherent absorbent pad rapidly wicks exudate. Easy to apply - Conforms to body contours and flexes with movement or swelling.
This item is not returnable. View return policy
Catalog No. NC1682429 Supplier McKesson General Medical Supplier No. 318721
3M Tegaderm +Pad Film Dressing with Non-Adherent Pad is an all-in-one dressing that provides a sterile, waterproof, viral and bacterial barrier. These dressings consist of a non-adherent absorbent pad bonded to a thin film transparent dressing. Practical for patients – Waterproof film lets patients bathe or shower without removing dressing. Enhances patient comfort – Unique, non-adherent pad will not stick to the wound, allowing normal wound healing with less pain and trauma. Unique, non-adherent absorbent pad rapidly wicks exudate. Easy to apply - Conforms to body contours and flexes with movement or swelling.