Learn More
RS Hughes Inc TGC Workgear Hi-Vis Orange Nitrile Diposable Glove - 2XL, 100/pk
SDP
Catalog No. NC1307993
Encompass_Preferred
The 2XL TGC Orange nitrile disposable glove provides comfort for extended wear while working with strong cleaners, acids, alkalis, solvents, biohazards, hydraulic fluids, oils and paint thinners. It's latex free, vinyl free and MBT free. Additionally, the TGC Orange glove features a beaded cuff, high-visibility orange color, and textured fingers and palm.
This item is not returnable. View return policy
Catalog No. NC1307993 Supplier RS Hughes Inc Supplier No. 34809800018
The 2XL TGC Orange nitrile disposable glove provides comfort for extended wear while working with strong cleaners, acids, alkalis, solvents, biohazards, hydraulic fluids, oils and paint thinners. It's latex free, vinyl free and MBT free. Additionally, the TGC Orange glove features a beaded cuff, high-visibility orange color, and textured fingers and palm.