Learn More
RS Hughes Inc TGC Workgear Hi-Vis Orange Nitrile Diposable Glove - Large, 100/pk
SDP
Catalog No. NC1307984
Encompass_Preferred
The TGC Work Gear Hi-Vis Orange disposable nitrile glove is highly resistant to a number of chemicals and liquids making it an excellent choice for industries and areas such as aviation, painting, laboratories, marine craft, petrol & diesel environments as well as commercial & industrial cleaning. Constructed from 6 mil Nitrile (Carboxylated Butadiene Acrylonitrile). The TGC Orange glove features a beaded cuff and textured palms and fingers. It is an excellent choice for comfort and extended wear. Completely latex, vinyl and MBT free.
This item is not returnable. View return policy
Catalog No. NC1307984 Supplier RS Hughes Inc Supplier No. 34809800016
The TGC Work Gear Hi-Vis Orange disposable nitrile glove is highly resistant to a number of chemicals and liquids making it an excellent choice for industries and areas such as aviation, painting, laboratories, marine craft, petrol & diesel environments as well as commercial & industrial cleaning. Constructed from 6 mil Nitrile (Carboxylated Butadiene Acrylonitrile). The TGC Orange glove features a beaded cuff and textured palms and fingers. It is an excellent choice for comfort and extended wear. Completely latex, vinyl and MBT free.