Learn More
RS Hughes Inc TGC Workgear Hi-Vis Orange Nitrile Disposable Glove -Medium, 100/pk
SDP
Catalog No. NC1307977
Encompass_Preferred
Need a high performance disposable glove with high resistance to chemicals and liquids such as cleaners, acids, alkalis, solvents, diesel, oils or petrol? If so, try the TGC Orange 6 mil nitrile glove. Constructed of Nitrile (Carboxylated Butadiene Acrylonitrile), this glove is latex free, MBT free and vinyl free. The nitrile material provides exceptional comfort for extended wear. The TGC Orange is an excellent choice within many industries and for a large variety of applications. The TGC Orange features a beaded cuff, textured finger and palm, and are ambidextrous.
This item is not returnable. View return policy
Catalog No. NC1307977 Supplier RS Hughes Inc Supplier No. 34809800015
Need a high performance disposable glove with high resistance to chemicals and liquids such as cleaners, acids, alkalis, solvents, diesel, oils or petrol? If so, try the TGC Orange 6 mil nitrile glove. Constructed of Nitrile (Carboxylated Butadiene Acrylonitrile), this glove is latex free, MBT free and vinyl free. The nitrile material provides exceptional comfort for extended wear. The TGC Orange is an excellent choice within many industries and for a large variety of applications. The TGC Orange features a beaded cuff, textured finger and palm, and are ambidextrous.