Learn More
RS Hughes Inc TGC Workgear Hi-Vis Orange Nitrile Diposable Glove - Extra Large, 100/pk
SDP
Catalog No. NC1307988
Encompass_Preferred
The XL Glove Company's TGC Hi-Vis Orange nitrile disposable glove is latex, vinyl and MBT free. It comes with a beaded cuff and has textured fingers and palm. The TGC Work Gear Orange glove offers high resistance to strong cleaners, acids, alkalis, solvents, biohazards, petrol, diesel, hydraulic fluids, oils and paint thinner. The 6 mil TGC Orange glove is constructed from Nitrile (Carboxylated Butadiene Acrylonitrile).
This item is not returnable. View return policy
Catalog No. NC1307988 Supplier RS Hughes Inc Supplier No. 34809800017
The XL Glove Company's TGC Hi-Vis Orange nitrile disposable glove is latex, vinyl and MBT free. It comes with a beaded cuff and has textured fingers and palm. The TGC Work Gear Orange glove offers high resistance to strong cleaners, acids, alkalis, solvents, biohazards, petrol, diesel, hydraulic fluids, oils and paint thinner. The 6 mil TGC Orange glove is constructed from Nitrile (Carboxylated Butadiene Acrylonitrile).