Learn More
Cayman Chemical Tigecycline, Purity: ≥98%, CAS: 220620-09-7, 5 mg
SDP
Catalog No. 502972262
Encompass_Preferred
A glycylcycline antibiotic; binds to the bacterial 30S ribosome, blocking the entry of transfer RNA, which halts protein synthesis and inhibits bacterial growth; active against a panel of 1,924 European clinical bacterial isolates (MICs = C. difficile cytotoxin activity and spore formation in cecum and colon in a mouse model of C. difficile infection at 6.25 mg/kg twice daily for 5 days
Catalog No. 50-297-2262 Supplier Cayman Chemical Supplier No. 150265 MG
A glycylcycline antibiotic; binds to the bacterial 30S ribosome, blocking the entry of transfer RNA, which halts protein synthesis and inhibits bacterial growth; active against a panel of 1,924 European clinical bacterial isolates (MICs = C. difficile cytotoxin activity and spore formation in cecum and colon in a mouse model of C. difficile infection at 6.25 mg/kg twice daily for 5 days