Learn More
Cayman Chemical TGF-1 LAPhumnrecombIn 500ug
SDP
Catalog No. 502943074
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged TGF-β1 LAP expressed in HEK293 cells • Amino acids: 1-278 (full length) • MW: 29.9 kDa
Catalog No. 50-294-3074 Supplier Cayman Chemical Supplier No. 42003500 UG
Source: Recombinant human C-terminal His-tagged TGF-β1 LAP expressed in HEK293 cells • Amino acids: 1-278 (full length) • MW: 29.9 kDa