Learn More
Cayman Chemical TGF-1 LAPhumnrecombIn 50ug
SDP
Catalog No. 502943073
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged TGF-β1 LAP expressed in HEK293 cells • Amino acids: 1-278 (full length) • MW: 29.9 kDa
Catalog No. 50-294-3073 Supplier Cayman Chemical Supplier No. 4200350 UG
Source: Recombinant human C-terminal His-tagged TGF-β1 LAP expressed in HEK293 cells • Amino acids: 1-278 (full length) • MW: 29.9 kDa