Learn More
Medchemexpress LLC TGF alpha/TGFA Mous 10ug | 10ug
SDP
Catalog No. 5000187672
Encompass_Preferred
TGF /TGFA is a mitogenic peptide that binds to the EGF receptor (EGFR) and acts synergistically with TGF to achieve anchorage-independent cell proliferation in soft agar It interacts with SDCBP and the SNTA1 PDZ domain which is critical for cell surface localization TGF alpha/TGFA Protein Mouse (HEK293 Fc) is the recombinant mouse-derived TGF alpha/TGFA protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-87672 Supplier Medchemexpress LLC Supplier No. HYP7785710UG
TGF /TGFA is a mitogenic peptide that binds to the EGF receptor (EGFR) and acts synergistically with TGF to achieve anchorage-independent cell proliferation in soft agar It interacts with SDCBP and the SNTA1 PDZ domain which is critical for cell surface localization TGF alpha/TGFA Protein Mouse (HEK293 Fc) is the recombinant mouse-derived TGF alpha/TGFA protein expressed by HEK293 with N-hFc labeled tag