Learn More
Santa Cruz Biotechnology TGFα Antibody (D-6): sc-374433, 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC374433
Encompass
TGFα Antibody (D-6) is a mouse monoclonal IgG1 κ TGFα antibody, cited in 8 publications, provided at 200 µg/mlspecific for an epitope mapping between amino acids 63-89 at the C-terminus of TGFα of human originTGF alpha Antibody (D-6) is recommended for detection of presursor and mature TGFα and BTC of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA; also reactive with additional species, including and equine, canine, bovine and porcine
Catalog No. NC3823846