Learn More
Eurofins Discoverx Recombinant Human TGF-β1 (HEK 293 derived), 2 µg. (92-1222)
Supplier:  Eurofins Discoverx 921222
Encompass
Transforming growth factor beta 1 (TGF-β1) is a cytokine peptide responsible for controlling proliferation, differentiation and other functions in cells. An agonist of TGFBR1, it is also involved in hematopoiesis, wound healing and immune system control, endothelial differentiation, chemotaxis, and the proliferation and differentiation of osteoblasts. Synthesized in many cell types, including most leukocytes, and acts at many receptors. Provided as 2μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9745