Learn More
Sino Biological Recombinant Human TGF beta 1 Protein (Latent, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10804H08HB100
Encompass_Preferred
A DNA sequence encoding the full length of human TGFβ1 (NP_000651.3) (Met1-Ser390) was fused with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6207