Learn More
Sino Biological Recombinant Mouse TGF beta 1 Protein (Latent, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50698M08HB20
Encompass_Preferred
A DNA sequence encoding the mouse Latent TGFB1 (P04202) (Met1-Ser390) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6076