Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Ca 2ug
SDP
Catalog No. 5000180566
Encompass_Preferred
TGFB1 proprotein is the precursor of latency-associated peptide (LAP) and active transforming growth factor Beta-1 (TGF- -1) chain which maintains TGF- -1 latency in the extracellular matrix TGFB1 binds non-covalently to TGF- -1 and interacts with environmental molecules (LTBP1 LRRC32/GARP LRRC33/NRROS) to regulate TGF- -1 activation TGF beta 1/TGFB1 Protein Canine (HEK293 His) is the recombinant canine-derived TGF beta 1/TGFB1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80566 Supplier Medchemexpress LLC Supplier No. HYP736172UG
TGFB1 proprotein is the precursor of latency-associated peptide (LAP) and active transforming growth factor Beta-1 (TGF- -1) chain which maintains TGF- -1 latency in the extracellular matrix TGFB1 binds non-covalently to TGF- -1 and interacts with environmental molecules (LTBP1 LRRC32/GARP LRRC33/NRROS) to regulate TGF- -1 activation TGF beta 1/TGFB1 Protein Canine (HEK293 His) is the recombinant canine-derived TGF beta 1/TGFB1 protein expressed by HEK293 with C-His labeled tag