Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Hu 100ug
SDP
Supplier:  Medchemexpress LLC HYP71056100UG
Encompass_Preferred
Recombinant human transforming growth factor beta-1 (TGF-β1) is a biotinylated, N-Avi-tagged protein expressed in HEK293 cells and supplied as a defined recombinant fragment (A279-S390). It is intended for research applications such as cell signaling studies, differentiation assays, and workflows that require a biotinylated human cytokine for capture or detection.
  • Biotinylated for streptavidin-based detection and capture.
  • Produced in HEK293 cells to retain human-like post-translational modifications.
  • Defined recombinant fragment corresponding to A279-S390.
  • Suitable for cell signaling, differentiation, and assay development.
  • Tagged format facilitates immobilization and assay integration.
Catalog No. 50-001-96742