Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Hu 100ug
SDP
Catalog No. 5000185515
Encompass_Preferred
Recombinant human transforming growth factor-beta 1 (TGF-β1) produced in HEK293 cells and supplied lyophilized for use in cell signaling, proliferation, and functional assays. The protein corresponds to residues A279-S390, exhibits high specific activity in bioassays, and is provided with low endotoxin to support cell-based applications.
  • Contains 100 μg of lyophilized protein for experimental use.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • High specific activity (ED50 ≈ 0.0964 ng/mL in Mv-1-lu cell assay).
  • Low endotoxin level <1 EU/μg, suitable for cell culture.
  • Lyophilized formulation for improved stability and shipping.
  • Store long term at -20°C; aliquot and freeze at -80°C for extended storage.
Catalog No. 50-001-85515 Supplier Medchemexpress LLC Supplier No. HYP70543100UG
Recombinant human transforming growth factor-beta 1 (TGF-β1) produced in HEK293 cells and supplied lyophilized for use in cell signaling, proliferation, and functional assays. The protein corresponds to residues A279-S390, exhibits high specific activity in bioassays, and is provided with low endotoxin to support cell-based applications.
  • Contains 100 μg of lyophilized protein for experimental use.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • High specific activity (ED50 ≈ 0.0964 ng/mL in Mv-1-lu cell assay).
  • Low endotoxin level <1 EU/μg, suitable for cell culture.
  • Lyophilized formulation for improved stability and shipping.
  • Store long term at -20°C; aliquot and freeze at -80°C for extended storage.